Mercury(II) acetate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12478-14
A12478-22
A12478-36
CAAAA12478-14EA
55.62
CAD
CAAAA12478-14
CAAAA12478-22
CAAAA12478-36
Mercury(II) acetate 98+%
Mercury (II) acetate
Purity: 98+%
CAS No.: 1600-27-7
Molecular Formula: C4H6HgO4
Formula Weight: 318.68
Melting Point: 179-182°
Beilstein Reference: 3563831
Merck Reference: 14,5873
UN No.: 1629
MDL No.: MFCD00012165
Formula:
(H₃CCOO)₂Hg Melting Pt: 179…182 °C Density: 3.28 Storage Temperature: Ambient |
MDL Number:
MFCD00012165 CAS Number: 1600-27-7 EINECS: 216-491-1 UN: 1629 ADR: 6.1,II Merck Index: 14,5873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...