Diethyl carbonate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12477-0D
A12477-0F
A12477-AP
CAAAA12477-0DEA
709.42
CAD
CAAAA12477-0D
CAAAA12477-0F
CAAAA12477-AP
Diethyl carbonate 99+%
Diethyl carbonate
Formula:
C₅H₁₀O₃ Boiling Pt: 126…128 °C Melting Pt: –43 °C Density: 0.975 Flash Pt: 31 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009107 CAS Number: 105-58-8 EINECS: 203-311-1 UN: 2366 ADR: 3,III Merck Index: 14,3780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...