5-(Hydroxymethyl)-2-furaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12475-03
A12475-06
A12475-14
CAAAA12475-03EA
77.1
CAD
CAAAA12475-03
CAAAA12475-06
CAAAA12475-14
5-(Hydroxymethyl)-2-furaldehyde 97%
5-(Hydroxymethyl)-2-furaldehyde
Purity: 97%
CAS No.: 67-47-0
Molecular Formula: C6H6O3
Formula Weight: 126.11
Storage Temperature: 2°C to 8°C
Flash Point: 79°(174°F)
Refractive Index: 1.5620
Melting Point: 32-35°
Boiling Point: 114-116°/1mm
Beilstein Reference: 110889
Merck Reference: 14,4832
MDL No.: MFCD00003234
Formula:
C₆H₆O₃ Boiling Pt: 114…116 °C/1 mm Melting Pt: 32…35 °C Density: 1.206 Flash Pt: 79 °C(174 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003234 CAS Number: 67-47-0 EINECS: 200-654-9 Merck Index: 14,4832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...