(2,4-Dichlorophenoxy)acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4-D
A12467-0B
A12467-30
CAAAA12467-0BEA
611.46
CAD
CAAAA12467-0B
CAAAA12467-30
(2,4-Dichlorophenoxy)acetic acid 98%
(2,4-Dichlorophenoxy)acetic acid
Purity: 98%
CAS No.: 94-75-7
Molecular Formula: C8H6Cl2O3
Formula Weight: 221.04
Odor: Odorless
Melting Point: 136-140°
Boiling Point: 160°/0.4mm
Beilstein Reference: 1214242
Merck Reference: 14,2796
MDL No.: MFCD00004300
Formula:
(C₆H₃Cl₂)OCH₂COOH MW: 221.04 g/mol Boiling Pt: 346 Melting Pt: 136…140 °C Density: 1.42 Storage Temperature: Ambient |
MDL Number:
MFCD00004300 CAS Number: 94-75-7 EINECS: 202-361-1 Merck Index: 14,2796 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...