3,4-Diaminopyridine 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12465-03 A12465-06 A12465-14
CAAAA12465-03EA 71.89 CAD
CAAAA12465-03 CAAAA12465-06 CAAAA12465-14
3,4-Diaminopyridine 98+%
3,4-Diaminopyridine

Purity: 98+% CAS No.: 54-96-6 Molecular Formula: C5H7N3 Formula Weight: 109.13 Flash Point: 240°(464°F) Odor: Unpleasant Beilstein Reference: 110232 Merck Reference: 14,2986 UN No.: 2811 MDL No.: MFCD00006401

Formula: C₅H₇N₃
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 217…220 °C
Flash Pt: 240 °C (464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006401
CAS Number: 54-96-6
EINECS: 200-220-9
UN: 2811
ADR: 6.1,III
Merck Index: 14,2986

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR