4-Nitroimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole

A12458-14 A12458-22
CAAAA12458-06EA 71.56 CAD
CAAAA12458-06 CAAAA12458-14 CAAAA12458-22
4-Nitroimidazole 97%
4-Nitroimidazole

Purity: 97% CAS No.: 3034-38-6 Molecular Formula: C3H3N3O2 Formula Weight: 113.08 Flash Point: 200°(392°F) Melting Point: ca 303° dec. Beilstein Reference: 2815 MDL No.: MFCD00005196

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: ∼ 303 °C (dec.)
Flash Pt: >200 °C (392 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005196
CAS Number: 3034-38-6
EINECS: 221-224-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR