Triphenylphosphine oxide 99%

Supplier: Thermo Scientific Chemicals
Warning

A12455-14 A12455-22 A12455-36
CAAAA12455-14EA 41.4 CAD
CAAAA12455-14 CAAAA12455-22 CAAAA12455-36
Triphenylphosphine oxide 99%
Triphenylphosphine oxide

Purity: 99% CAS No.: 791-28-6 Molecular Formula: C18H15OP Formula Weight: 278.29 Flash Point: 180°(356°F) Melting Point: 153-159° Boiling Point: 360° Beilstein Reference: 745854 MDL No.: MFCD00002080

Formula: C₁₈H₁₅OP
MW: 278.29 g/mol
Boiling Pt: 360 °C
Melting Pt: 153…159 °C
Density: 1.200
Flash Pt: 180 °C(356 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002080
CAS Number: 791-28-6
EINECS: 212-338-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR