3'-Bromoacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12454-14
A12454-22
A12454-36
CAAAA12454-14EA
81.37
CAD
CAAAA12454-14
CAAAA12454-22
CAAAA12454-36
3'-Bromoacetophenone 98+%
3'-Bromoacetophenone
Purity: 98+%
CAS No.: 2142-63-4
Molecular Formula: C8H7BrO
Formula Weight: 199.05
Flash Point: 114°(237°F)
Refractive Index: 1.5750
Melting Point: 7-8°
Boiling Point: 79-81°/2mm
Beilstein Reference: 507599
MDL No.: MFCD00000083
Formula:
C₈H₇BrO Boiling Pt: 79…81 °C/2 mm Melting Pt: 7…8 °C Density: 1.498 Flash Pt: 114 °C(237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000083 CAS Number: 2142-63-4 EINECS: 218-396-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...