5-Nitrouracil 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nitrouracil, 5-Nitropyrimidine-2,4(1H,3H)-dione, 2,4-Dihydroxy-5-nitropyrimidine

A12448-14 A12448-22
CAAAA12448-14EA 73.63 CAD
CAAAA12448-14 CAAAA12448-22 CAAAA12448-36
5-Nitrouracil 98+%
5-Nitrouracil

Purity: 98+% CAS No.: 611-08-5 Molecular Formula: C4H3N3O4 Formula Weight: 157.09 Melting Point: ca 300° Beilstein Reference: 10410

Formula: C₄H₃N₃O₄
MW: 157.09 g/mol
Melting Pt: ∼ 300 °C
Density: 1.807 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006021
CAS Number: 611-08-5
EINECS: 210-250-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR