5-Amino-3-methylisoxazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A12446-03 A12446-06 A12446-14
CAAAAA12446-03EA 72.21 CAD
CAAAAA12446-03 CAAAA12446-06 CAAAA12446-14
5-Amino-3-methylisoxazole 98%
5-Amino-3-methylisoxazole

Purity: 98+% CAS No.: 14678-02-5 Molecular Formula: C4H6N2O Formula Weight: 98.11 Melting Point: 81-85° Beilstein Reference: 107906

Formula: C₄H₆N₂O
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00003151
CAS Number: 14678-02-5
EINECS: 238-719-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR