2,5-Diiodothiophene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12443-06
A12443-14
CAAAAA12443-22EA
2762.6
CAD
CAAAAA12443-22
CAAAA12443-06
CAAAA12443-14
2,5-Diiodothiophene 99%
2,5-Diiodothiophene
Purity: 99%
CAS No.: 625-88-7
Molecular Formula: C4H2I2S
Formula Weight: 335.93
Storage Temperature: 2°C to 8°C
Melting Point: 39-43°
Boiling Point: 139-140°/15mm
Beilstein Reference: 109915
MDL No.: MFCD00014525
Formula:
C₄H₂I₂S Boiling Pt: 139…140 °C/15 mm Melting Pt: 39…43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014525 CAS Number: 625-88-7 EINECS: 210-915-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...