Homovanillic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12441-14
A12441-03
A12441-06
CAAAAA12441-14EA
1161.3
CAD
CAAAAA12441-14
CAAAA12441-03
CAAAA12441-06
Homovanillic acid 98+%
Homovanillic acid
Fluorimetric reagent and major catecholamine metabolite
Purity: 98+%
CAS No.: 306-08-1
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 140-145°
Beilstein Reference: 2213447
Merck Reference: 14,4740
MDL No.: MFCD00004350
Formula:
(C₆H₃(OH)(OCH₃))CH₂COOH Boiling Pt: 339 °C (1013 hPa) Melting Pt: 140…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004350 CAS Number: 306-08-1 EINECS: 206-176-7 Merck Index: 14,4740 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...