1-Octyne 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12440-09
A12440-14
A12440-22
CAAAAA12440-09EA
55.62
CAD
CAAAAA12440-09
CAAAA12440-14
CAAAA12440-22
1-Octyne 98%
1-Octyne
Purity: 98%
CAS No.: 629-05-0
Molecular Formula: C8H14
Formula Weight: 110.20
Flash Point: 17°(62°F)
Physical Form: Liquid
Density: 0.746
Refractive Index: 1.4160
Appearance: Colorless
Melting Point: -60°
Boiling Point: 123-126°
Beilstein Reference: 1734494
UN No.: 3295
MDL No.: MFCD00009546
Formula:
C₈H₁₄ Boiling Pt: 123…126 °C Melting Pt: –60 °C Density: 0.746 Flash Pt: 17 °C (62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009546 CAS Number: 629-05-0 EINECS: 211-069-3 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...