1,6-Hexanediol 97%
Supplier: Thermo Scientific Chemicals
A12439-30
A12439-0E
A12439-36
CAAAAA12439-30EA
46.45
CAD
CAAAAA12439-30
CAAAA12439-0E
CAAAA12439-36
1,6-Hexanediol 97%
1,6-Hexanediol
Purity: 97%
CAS No.: 629-11-8
Molecular Formula: HO(CH2)6OH
Formula Weight: 118.18
Flash Point: 147°(296°F)
Physical Form: Cryst solid
Density: 1.120
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 40-43°
Boiling Point: 248-250°
Beilstein Reference: 1633461
Merck Reference: 14,4690
MDL No.: MFCD00002985
Formula:
C₆H₁₄O₂ Boiling Pt: 248…250 °C Melting Pt: 40…43 °C Density: 1.120 Flash Pt: 147 °C(296 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002985 CAS Number: 629-11-8 EINECS: 211-074-0 Merck Index: 14,4690 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...