4-Phenylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A12438-14 A12438-22 A12438-36
CAAAA12438-14EA 71.1 CAD
CAAAA12438-14 CAAAA12438-22 CAAAA12438-36
4-Phenylbenzoic acid 98%
4-Phenylbenzoic acid

Purity: 98% CAS No.: 92-92-2 Molecular Formula: C13H10O2 Formula Weight: 198.22 Odor: Odorless Melting Point: 223-227° Beilstein Reference: 973519 MDL No.: MFCD00002553

Formula: C₁₃H₁₀O₂
Boiling Pt: 373 °C (1013 hPa)
Melting Pt: 223…227 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002553
CAS Number: 92-92-2
EINECS: 202-203-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR