2,7-Dibromofluorene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,7-Dibromo-9H-fluorene

A12437-09 A12437-22 A12437-14
CAAAAA12437-09EA 76.95 CAD
CAAAAA12437-09 CAAAAA12437-22 CAAAA12437-14
2,7-Dibromofluorene 98%
2,7-Dibromofluorene

Purity: 98% CAS No.: 16433-88-8 Molecular Formula: C13H8Br2 Formula Weight: 324.03 Melting Point: 164-167° Beilstein Reference: 2049205

Formula: C₁₃H₈Br₂
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00019048
CAS Number: 16433-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR