1,5-Dichloropentane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pentamethylendichloride
A12433-22
A12433-36
CAAAA12433-22EA
164.32
CAD
CAAAA12433-22
CAAAA12433-36
1,5-Dichloropentane 98%
1,5-Dichloropentane
Purity: 98%
CAS No.: 628-76-2
Molecular Formula: C5H10Cl2
Formula Weight: 141.04
Flash Point: 26°(78°F)
Refractive Index: 1.4570
Melting Point: -72°
Boiling Point: 182°
Beilstein Reference: 1732340
UN No.: 1152
MDL No.: MFCD00001017
Formula:
ClH₂C(CH₂)₃CH₂Cl MW: 141.04 g/mol Boiling Pt: 63 Melting Pt: -72.8 Density: 1.103 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001017 CAS Number: 628-76-2 EINECS: 211-053-6 UN: 1152 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...