Chlorodifluoroacetic anhydride 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12432-09 A12432-18
CAAAA12432-09EA 109.81 CAD
CAAAA12432-09 CAAAA12432-18 CAAAA12432-30
Chlorodifluoroacetic anhydride 98+%
Chlorodifluoroacetic anhydride

Purity: 98+% CAS No.: 2834-23-3 Molecular Formula: (ClCF2CO)2O Formula Weight: 242.94 Refractive Index: 1.3470 Boiling Point: 92-93° UN No.: 3265 MDL No.: MFCD00000774

Formula: C₄Cl₂F₄O₃
Boiling Pt: 92…93 °C
Density: 1.393
Storage Temperature: Ambient
MDL Number: MFCD00000774
CAS Number: 2834-23-3
EINECS: 220-603-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR