Pentafluorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzoyl chloride, Pentafluorobenzoylchloride, PFBC

A12420-06 A12420-14 A12420-22
CAAAA12420-06EA 105.39 CAD
CAAAA12420-06 CAAAA12420-14 CAAAA12420-22
Pentafluorobenzoyl chloride 98%
Pentafluorobenzoyl chloride

Purity: 98% CAS No.: 2251-50-5 Molecular Formula: C7ClF5O Formula Weight: 230.52 Refractive Index: 1.4535 Boiling Point: 158-159° UN No.: 3265 MDL No.: MFCD00000657

Formula: C₇ClF₅O
Boiling Pt: 158…159 °C
Density: 1.668
Storage Temperature: Ambient
MDL Number: MFCD00000657
CAS Number: 2251-50-5
EINECS: 218-842-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR