2-Fluoro-5-nitrobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12417-03
A12417-06
CAAAA12417-03EA
86.9
CAD
CAAAA12417-03
CAAAA12417-06
2-Fluoro-5-nitrobenzonitrile 98+%
2-Fluoro-5-nitrobenzonitrile
Purity: 98+%
CAS No.: 17417-09-3
Molecular Formula: C7H3FN2O2
Formula Weight: 166.11
Melting Point: 70-72°
Beilstein Reference: 2048720
UN No.: 3439
MDL No.: MFCD00042299
Formula:
C₇H₃FN₂O₂ Melting Pt: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042299 CAS Number: 17417-09-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...