2,4-Difluorophenyl isocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12415-06
A12415-14
A12415-22
CAAAA12415-06EA
72.05
CAD
CAAAA12415-06
CAAAA12415-14
CAAAA12415-22
2,4-Difluorophenyl isocyanate 98+%
2,4-Difluorophenyl isocyanate
Purity: 98+%
CAS No.: 59025-55-7
Molecular Formula: C7H3F2NO
Formula Weight: 155.11
Flash Point: 50°(122°F)
Refractive Index: 1.4920
Boiling Point: 64-66°/10mm
Beilstein Reference: 2831577
UN No.: 3080
MDL No.: MFCD00001997
Formula:
C₇H₃F₂NO Boiling Pt: 64…66 °C/10 mm Density: 1.310 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001997 CAS Number: 59025-55-7 UN: 2478 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...