2-Bromobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-Bromophenyl)methanamine

A12412-06 A12412-14
CAAAA12412-06EA 134.77 CAD
CAAAA12412-06 CAAAA12412-14 CAAAA12412-22
2-Bromobenzylamine 97%
2-Bromobenzylamine

Purity: 98% CAS No.: 3959-05-5 Molecular Formula: C7H8BrN Formula Weight: 186.06 Odor: Amine-like Boiling Point: 118-119°/9mm UN No.: 2735 MDL No.: MFCD00025572

Formula: C₇H₈BrN
MW: 186.05
Boiling Pt: 118…119 °C (9 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00025572
CAS Number: 3959-05-5
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR