1-Fluoro-2-iodobenzene 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
A12411-14
A12411-22
A12411-36
CAAAA12411-14EA
58.46
CAD
CAAAA12411-14
CAAAA12411-22
CAAAA12411-36
1-Fluoro-2-iodobenzene 99% stabilized
1-Fluoro-2-iodobenzene
Purity: 99%
CAS No.: 348-52-7
Molecular Formula: C6H4FI
Formula Weight: 222.00
Flash Point: 71°(159°F)
Refractive Index: 1.5900
Melting Point: -43° to -40°
Boiling Point: 188-189°
Beilstein Reference: 2039770
MDL No.: MFCD00001031
Formula:
C₆H₄FI Boiling Pt: 188…189 °C Melting Pt: –43 °C…–40 °C Density: 1.903 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001031 CAS Number: 348-52-7 EINECS: 206-477-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...