Iodoacetic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12410-14
A12410-22
CAAAA12410-14EA
146.47
CAD
CAAAA12410-14
CAAAA12410-22
Iodoacetic acid 97+%
Iodoacetic acid
As a reagent for the modification of sulfhydryl groups
Purity: 97+%
CAS No.: 64-69-7
Molecular Formula: ICH2CO2H
Formula Weight: 185.95
Storage Temperature: 2°C to 8°C
Odor: Pungent
Boiling Point: 208° dec.
Beilstein Reference: 1739079
Merck Reference: 14,5025
UN No.: 2923
MDL No.: MFCD00002685
Formula:
C₂H₃IO₂ Boiling Pt: 208 °C (dec.) Melting Pt: 79…82 °C Density: 2.2694 g/cm³ (85 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002685 CAS Number: 64-69-7 EINECS: 200-590-1 UN: 2923 ADR: 8,III Merck Index: 14,5025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...