Benzoic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl benzoate

A12404-0B A12404-0I A12404-30
CAAAA12404-0BEA 92.9 CAD
CAAAA12404-0B CAAAA12404-0I CAAAA12404-30
Benzoic acid methyl ester 99%
Benzoic acid methyl ester

Purity: 99% CAS No.: 93-58-3 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 82°(179°F) Physical Form: Liquid Density: 1.094 Refractive Index: 1.5170 Appearance: Colorless Melting Point: -12° Boiling Point: 198-200° Beilstein Reference: 1072099 Merck Reference: 14,6024 MDL No.: MFCD00008421

Formula: C₆H₅COOCH₃
Boiling Pt: 198…200 °C
Melting Pt: –12 °C
Density: 1.094
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 14,6024
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR