Taurine 99%

Supplier: Thermo Scientific Chemicals

A12403-0E A12403-22 A12403-36
CAAAA12403-0EEA 785.26 CAD
CAAAA12403-0E CAAAA12403-22 CAAAA12403-36
Taurine 99%
Taurine

Purity: 99% CAS No.: 107-35-7 Molecular Formula: H2NCH2CH2SO3H Formula Weight: 125.15 Physical Form: Crystals or cryst powder Appearance: White Melting Point: 300° Beilstein Reference: 1751215 Merck Reference: 14,9074 MDL No.: MFCD00008197

Formula: C₂H₇NO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 14,9074

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR