(S)-(-)-Nicotine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine

A12398-14 A12398-22
CAAAA12398-14EA 118.82 CAD
CAAAA12398-14 CAAAA12398-22
(S)-(-)-Nicotine 99%
(S)-(-)-Nicotine

Purity: 99% CAS No.: 54-11-5 Molecular Formula: C10H14N2 Formula Weight: 162.24 Flash Point: 101°(213°F) Refractive Index: 1.5265 Odor: Characteristic pH: 10.2 Melting Point: -79° Boiling Point: 245-248° Beilstein Reference: 82109 Merck Reference: 14,6524 Optical Rotation: -168° (neat) UN No.: 1654 MDL No.: MFCD00006369

Formula: C₁₀H₁₄N₂
Boiling Pt: 245…248 °C
Melting Pt: –79 °C
Density: 1.017
Flash Pt: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1,II
Merck Index: 14,6524

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR