2-Aminopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Pyridylamine, Pyridin-2-amine

A12374-14 A12374-0B A12374-30
CAAAAA12374-14EA 52.46 CAD
CAAAAA12374-14 CAAAA12374-0B CAAAA12374-30
2-Aminopyridine 99%
2-Aminopyridine

Purity: 99% CAS No.: 504-29-0 Molecular Formula: C5H6N2 Formula Weight: 94.12 Flash Point: 92°(197°F) Melting Point: 56-60° Boiling Point: 209-211° Beilstein Reference: 105785 Merck Reference: 14,473 UN No.: 2671 MDL No.: MFCD00006312

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 209…211 °C
Melting Pt: 56…60 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II
Merck Index: 14,473

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR