4-Chlorobenzoylacetonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12364-22
A12364-06
A12364-14
CAAAAA12364-22EA
1221.34
CAD
CAAAAA12364-22
CAAAA12364-06
CAAAA12364-14
4-Chlorobenzoylacetonitrile 98%
4-Chlorobenzoylacetonitrile
Purity: 98%
CAS No.: 4640-66-8
Molecular Formula: C9H6ClNO
Formula Weight: 179.61
Melting Point: 128-132°
Beilstein Reference: 743368
Formula:
C₉H₆ClNO Melting Pt: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051625 CAS Number: 4640-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...