Diphenyl carbonate 99%

Supplier: Thermo Scientific Chemicals
Warning

A12362-0B A12362-0I A12362-30
CAAAA12362-0BEA 133.98 CAD
CAAAA12362-0B CAAAA12362-0I CAAAA12362-30
Diphenyl carbonate 99%
Diphenyl carbonate

Purity: 99% CAS No.: 102-09-0 Molecular Formula: C13H10O3 Formula Weight: 214.22 Flash Point: 168°(334°F) Odor: Mild Boiling Point: 301-302° Beilstein Reference: 1074863 Merck Reference: 14,3602 MDL No.: MFCD00003037

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 301
Melting Pt: 78
Density: 1.300
Flash Pt: 168 °C (334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003037
CAS Number: 102-09-0
EINECS: 203-005-8
UN: 3077
ADR: 9 III
Merck Index: 14,3602

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR