1,10-Dibromodecane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,10-Dibromodecan, Decane, 1,10-dibromo-

A12357-22 A12357-36
CAAAA12357-22EA 180.12 CAD
CAAAA12357-22 CAAAA12357-36
1,10-Dibromodecane 97%
1,10-Dibromodecane

Purity: 97% CAS No.: 4101-68-2 Molecular Formula: Br(CH2)10Br Formula Weight: 300.08 Flash Point: 110°(230°F) Refractive Index: 1.4930 Melting Point: 26-30° Boiling Point: 157-159°/14mm Beilstein Reference: 506156 MDL No.: MFCD00000222

Formula: BrH₂C(CH₂)₈CH₂Br
MW: 300.08 g/mol
Boiling Pt: 157…159 °C (14 mmHg)
Melting Pt: 26…30 °C
Density: 1.340
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000222
CAS Number: 4101-68-2
EINECS: 223-871-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR