2,5-Dimethylpyrazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12352-06
A12352-14
A12352-22
CAAAA12352-06EA
59.88
CAD
CAAAA12352-06
CAAAA12352-14
CAAAA12352-22
2,5-Dimethylpyrazine 99%
2,5-Dimethylpyrazine
Purity: 99%
CAS No.: 123-32-0
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 63°(145°F)
Refractive Index: 1.5 00
Odor: Pleasant
Melting Point: 15°
Boiling Point: 154-155°
Beilstein Reference: 107052
MDL No.: MFCD00006147
Formula:
C₆H₈N₂ Boiling Pt: 154…155 °C Melting Pt: 15 °C Density: 0.990 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006147 CAS Number: 123-32-0 EINECS: 204-618-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...