1-Iodobutane 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with copper
A12340-22
A12340-36
CAAAA12340-22EA
67.94
CAD
CAAAA12340-22
CAAAA12340-36
1-Iodobutane 99% stabilized
1-Iodobutane
Purity: 99%
CAS No.: 542-69-8
Molecular Formula: C4H9I
Formula Weight: 184.02
Flash Point: 33°(91°F)
Refractive Index: 1.4990
Melting Point: -103°
Boiling Point: 129-131°
Beilstein Reference: 1420755
Merck Reference: 14,1573
UN No.: 1993
MDL No.: MFCD00001098
Formula:
C₄H₉I Boiling Pt: 129…131 °C Melting Pt: –103 °C Density: 1.616 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001098 CAS Number: 542-69-8 EINECS: 208-824-4 UN: 2929 ADR: 6.1,II Merck Index: 14,1573 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...