2-Nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Nitrophenol

A12333-0B A12333-30
CAAAAA12333-0IEA 605.65 CAD
CAAAAA12333-0I CAAAA12333-0B CAAAA12333-30
2-Nitrophenol 98%
2-Nitrophenol

Purity: 98% CAS No.: 88-75-5 Molecular Formula: C6H5NO3 Formula Weight: 139.11 Flash Point: 108°(226°F) Odor: Phenol-like Boiling Point: 214-216° Beilstein Reference: 775403 Merck Reference: 14,6620 UN No.: 1663 MDL No.: MFCD00011688

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 214…216 °C
Melting Pt: 44…48 °C
Density: 1.26
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1,III
Merck Index: 14,6620

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR