4-Bromo-3-methylanisole 97%

Supplier: Thermo Scientific Chemicals
Warning

A12318-09 A12318-18 A12318-30
CAAAA12318-09EA 92.59 CAD
CAAAA12318-09 CAAAA12318-18 CAAAA12318-30
4-Bromo-3-methylanisole 97%
4-Bromo-3-methylanisole

Purity: 97% CAS No.: 27060-75-9 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 110°(230°F) Refractive Index: 1.5610 Boiling Point: 111-112°/15mm Beilstein Reference: 1933865 MDL No.: MFCD00060687

Formula: C₈H₉BrO
Boiling Pt: 111…112 °C (15 mmHg)
Density: 1.450
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR