2-Bromotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

A12315-14 A12315-22 A12315-36
CAAAA12315-14EA 80.26 CAD
CAAAA12315-14 CAAAA12315-22 CAAAA12315-36
2-Bromotoluene 99%
2-Bromotoluene

Purity: 99% CAS No.: 95-46-5 Molecular Formula: C7H7Br Formula Weight: 171.04 Flash Point: 78°(172°F) Refractive Index: 1.5560 Melting Point: -27° Boiling Point: 179-181° Beilstein Reference: 1904176 Merck Reference: 14,1439 MDL No.: MFCD00000068

Formula: C₇H₇Br
MW: 171.04 g/mol
Boiling Pt: 179…181 °C
Melting Pt: –27 °C
Density: 1.420
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000068
CAS Number: 95-46-5
EINECS: 202-421-7
UN: 3082
Merck Index: 14,1439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR