L(+)-Leucine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

A12311-0E A12311-22 A12311-36
CAAAA12311-0EEA 1422 CAD
CAAAA12311-0E CAAAA12311-22 CAAAA12311-36
L(+)-Leucine 99%
L(+)-Leucine

Purity: 99% CAS No.: 61-90-5 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Physical Form: Cryst powder Density: 1.293 Appearance: White Melting Point: 300° Beilstein Reference: 1721722 Merck Reference: 14,5451 Optical Rotation: +15° (c=2 in 5N HCl) MDL No.: MFCD00002617

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: >300 °C
Density: 1.293
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,5451

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR