97+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate

A12310-06 A12310-14 A12310-22
CAAAA12310-06EA 106.97 CAD
CAAAA12310-06 CAAAA12310-14 CAAAA12310-22
97+%
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP)

Substrate for alkaline and acid phosphatase determinations.


4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

Formula: C₆H₄NNa₂O₆P·6H₂O
MW: 371.144 g/mol
Melting Pt: 300
Storage Temperature: Refrigerator
MDL Number: MFCD00066288
CAS Number: 333338-18-4
EINECS: 224-246-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR