4-Ethoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
A12309-14
A12309-22
A12309-36
CAAAA12309-14EA
64.62
CAD
CAAAA12309-14
CAAAA12309-22
CAAAA12309-36
4-Ethoxybenzoic acid 98+%
4-Ethoxybenzoic acid
Purity: 98+%
CAS No.: 619-86-3
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 197-199°
Beilstein Reference: 2207349
Formula:
C₉H₁₀O₃ Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002545 CAS Number: 619-86-3 EINECS: 210-616-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...