Methyl-2,4,6-trihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A12305-06 A12305-14 A12305-22
CAAAA12305-06EA 95.75 CAD
CAAAA12305-06 CAAAA12305-14 CAAAA12305-22
Methyl-2,4,6-trihydroxybenzoate 98%
Methyl 2,4,6-trihydroxybenzoate

Purity: 98% CAS No.: 3147-39-5 Molecular Formula: C8H8O5 Formula Weight: 184.15 Melting Point: 175-179° Beilstein Reference: 2837819

Formula: C₈H₈O₅
Melting Pt: 175…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00013969
CAS Number: 3147-39-5
EINECS: 221-566-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR