β-Chloropropiophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloropropiophenone

A12290-22 A12290-09 A12290-18
CAAAAA12290-22EA 418.7 CAD
CAAAAA12290-22 CAAAA12290-09 CAAAA12290-18
β-Chloropropiophenone 96%
β-Chloropropiophenone

Purity: 96% CAS No.: 936-59-4 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 46-50° Boiling Point: 113-115°/4mm Beilstein Reference: 2043580 MDL No.: MFCD00000990

Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168.62 g/mol
Boiling Pt: 263
Melting Pt: 46…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR