Dibenzothiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12288-14
A12288-22
A12288-36
CAAAA12288-14EA
85.95
CAD
CAAAA12288-14
CAAAA12288-22
CAAAA12288-36
Dibenzothiophene 98%
Dibenzothiophene
Purity: 98%
CAS No.: 132-65-0
Molecular Formula: C12H8S
Formula Weight: 184.25
Flash Point: 170°(338°F)
Melting Point: 97-100°
Boiling Point: 332-333°
Beilstein Reference: 121101
UN No.: 3077
MDL No.: MFCD00004969
Formula:
C₁₂H₈S Boiling Pt: 332…333 °C Melting Pt: 97…100 °C Density: 1.1295 g/cm³ (125 °C) Flash Pt: 170 °C (338 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004969 CAS Number: 132-65-0 EINECS: 205-072-9 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...