2-Bromomesitylene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

A12277-22 A12277-36 A12277-18 A12277-30
CAAAAA12277-22EA 115.18 CAD
CAAAAA12277-22 CAAAAA12277-36 CAAAA12277-18 CAAAA12277-30
2-Bromomesitylene 99%
2-Bromomesitylene

Purity: 99% CAS No.: 576-83-0 Molecular Formula: C9H11Br Formula Weight: 199.10 Flash Point: 96°(204°F) Refractive Index: 1.5520 Melting Point: 2° Boiling Point: 224-226° Beilstein Reference: 1907245 MDL No.: MFCD00000073

Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 224…226 °C
Melting Pt: 2 °C
Density: 1.322
Flash Pt: 96 °C(204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS Number: 576-83-0
EINECS: 209-405-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR