3-Fluorophthalic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12275-06
A12275-14
CAAAA12275-06EA
164.32
CAD
CAAAA12275-06
CAAAA12275-14
3-Fluorophthalic anhydride 98%
3-Fluorophthalic anhydride
Purity: 98%
CAS No.: 652-39-1
Molecular Formula: C8H3FO3
Formula Weight: 166.11
Melting Point: 158-163°
Beilstein Reference: 1283580
Formula:
C₈H₃FO₃ Boiling Pt: 351 °C (1013 hPa) Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039696 CAS Number: 652-39-1 EINECS: 211-491-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...