2-Piperidone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12271-14
A12271-22
CAAAAA12271-36EA
1587.38
CAD
CAAAAA12271-36
CAAAA12271-14
CAAAA12271-22
2-Piperidone 98%
2-Piperidone
Purity: 98+%
CAS No.: 675-20-7
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 147°(296°F)
Melting Point: 35-39°
Boiling Point: 255-257°
Beilstein Reference: 106434
MDL No.: MFCD00006037
Formula:
C₅H₉NO Boiling Pt: 255…257 °C Melting Pt: 35…39 °C Density: 1.073 g/cm³ (40 °C) Flash Pt: 147 °C(296 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006037 CAS Number: 675-20-7 EINECS: 211-622-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...