Eucalyptol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane
A12269-AE
A12269-AP
CAAAA12269-AEEA
62.41
CAD
CAAAA12269-AE
CAAAA12269-AP
Eucalyptol 99%
Eucalyptol
Purity: 99%
CAS No.: 470-82-6
Molecular Formula: C10H18O
Formula Weight: 154.24
Flash Point: 49°(120°F)
Refractive Index: 1.4575
Melting Point: 1.0°
Boiling Point: 176-177°
Beilstein Reference: 105109
Merck Reference: 14,3895
UN No.: 3271
MDL No.: MFCD00167977
Formula:
C₁₀H₁₈O Boiling Pt: 176…177 °C Melting Pt: 1.0 °C Density: 0.923 Flash Pt: 49 °C (120 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00167977 CAS Number: 470-82-6 EINECS: 207-431-5 UN: 1993 ADR: 3,III Merck Index: 14,3895 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...