Benzhydrazide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzoylhydrazine, Benzoic acid hydrazide, Benzohydrazide, Benzoic hydrazide
A12268-14
A12268-22
A12268-36
CAAAAA12268-14EA
51.82
CAD
CAAAAA12268-14
CAAAA12268-22
CAAAA12268-36
Benzhydrazide 98%
Benzhydrazide
Purity: 98%
CAS No.: 613-94-5
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 113-117°
Beilstein Reference: 471797
Formula:
C₇H₈N₂O MW: 136.15 g/mol Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007596 CAS Number: 613-94-5 EINECS: 210-363-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...