Pyridine-2,6-dicarboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA
A12263-14
A12263-22
A12263-36
CAAAA12263-14EA
61.15
CAD
CAAAA12263-14
CAAAA12263-22
CAAAA12263-36
Pyridine-2,6-dicarboxylic acid 98%
Pyridine-2,6-dicarboxylic acid
Purity: 98%
CAS No.: 499-83-2
Molecular Formula: C7H5NO4
Formula Weight: 167.12
Flash Point: 188°(370°F)
Melting Point: ca 245° dec.
Beilstein Reference: 131629
MDL No.: MFCD00006299
Formula:
C₇H₅NO₄ MW: 167.12 g/mol Boiling Pt: 357 Melting Pt: ∼ 245 °C (dec.) Flash Pt: 188 °C (370 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006299 CAS Number: 499-83-2 EINECS: 207-894-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...