1-Bromo-3-ethylbenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A12258-03 A12258-06 A12258-14
CAAAA12258-03EA 42.98 CAD
CAAAA12258-03 CAAAA12258-06 CAAAA12258-14
1-Bromo-3-ethylbenzene 98%
1-Bromo-3-ethylbenzene

Purity: 98% CAS No.: 2725-82-8 Molecular Formula: C8H9Br Formula Weight: 185.06 Boiling Point: 76-78°/10mm MDL No.: MFCD00156128

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 76…78 °C (10 mmHg)
Density: 1,3493
Storage Temperature: Ambient
MDL Number: MFCD00156128
CAS Number: 2725-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR