N-(Hydroxymethyl)benzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

A12250-14 A12250-22 A12250-09 A12250-30
CAAAAA12250-14EA 111.39 CAD
CAAAAA12250-14 CAAAAA12250-22 CAAAA12250-09 CAAAA12250-30
N-(Hydroxymethyl)benzamide 98%
N-(Hydroxymethyl)benzamide

Purity: 98% CAS No.: 6282-02-6 Molecular Formula: C6H5CONHCH2OH Formula Weight: 151.17 Melting Point: 94-96° Beilstein Reference: 2045129

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00014415
CAS Number: 6282-02-6
EINECS: 228-495-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR